missing translation for 'onlineSavingsMsg'
Learn More

4'-Methylacetophenone, TRC

Product Code. 30481301
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481301 100 g 100g
30468550 25 g 25g
30469714 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30481301 Supplier Toronto Research Chemicals Supplier No. TRCM275305100G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-00-9
Molecular Formula C9 H10 O
Molecular Weight (g/mol) 134.18
Synonym Ethanone, 1-(4-methylphenyl)-, Acetophenone, 4'-methyl- (8CI), 1-(4-Methylphenyl)ethanone, 1-(4-Methylphenyl)-1-ethanone, 1-(4-Tolyl)ethanone, 1-(p-Tolyl)ethan-1-one, 1-Acetyl-4-methylbenzene, 1-p-Tolylethanone, 4-Acetyltoluene, 4-Methyl-1-acetylbenzene, 4-Methylhypnone, 4-Methylphenyl methyl ketone, 4'-Methylacetophenone, Melilotal, Methyl 4-methylphenyl ketone, Methyl p-tolyl ketone, NSC 9401, p-Acetyltoluene, p-Methylacetophenone, p-Tolyl methyl ketone
IUPAC Name 1-(4-methylphenyl)ethanone
SMILES CC(=O)c1ccc(C)cc1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C9H10O/c1-7-3-5-9(6-4-7)8(2)10/h3-6H,1-2H3
Formula Weight 134.0732
Chemical Name or Material 4'-Methylacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.