missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Methylamino)-1-(3-pyridyl-d4)-1-butanone 2HCl, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 4-(Methylamino)-1-(3-pyridyl-d4)-1-butanone Dihydrochloride |
| CAS | 764661-23-6 |
| Molecular Formula | C10 2H4 H10 N2 O . 2 Cl H |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C10H14N2O0.2 ClH/c1-11-6-3-5-10(13)9-4-2-7-12-8-9;;/h2,4,7-8,11 H,3,5-6H2,1H3;2*1 H/i2D,4 D,7 D,8 D;; |
| SMILES | Cl.Cl.[2 H]c1nc([2 H])c(C(=O)CCCNC)c([2 H])c1[2 H] |
| IUPAC Name | 4-(methylamino)-1-(2,4,5,6-tetradeuteriopyridin-3-yl)butan-1-one;dihydrochloride |
| Molecular Weight (g/mol) | 255.18 |
| Formula Weight | 254.0891 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction