missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzeneboronic acid, 98%

Product Code. 11440973
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440973 1 g 1g
11450973 5 g 5g
11460973 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440973 Supplier Thermo Scientific Alfa Aesar Supplier No. A13347.03
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 5720-05-8 | C7H9BO2 | 135.96 g/mol

4-Methylbenzeneboronic acid, is a commonly used chemical in Suzuki coupling reactions. It is also used as an intermediate in pharmaceutical industry

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylbenzeneboronic acid, is a commonly used chemical in Suzuki coupling reactions. It is also used as an intermediate in pharmaceutical industry

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5720-05-8

Specifications

Melting Point 246°C to 254°C
Molecular Formula C7H9BO2
MDL Number MFCD00039138
Quantity 1 g
Beilstein 2935970
Synonym 4-tolylboronic acid, p-tolylboronic acid, 4-methylphenyl boronic acid, p-tolueneboronic acid, 4-methylbenzeneboronic acid, p-methylbenzeneboronic acid, boronic acid, 4-methylphenyl, boronic acid, p-tolyl, 4-methylphenylboric acid, tolyl boronic acid
Solubility Information It is soluble in water.
InChI Key BIWQNIMLAISTBV-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)B(O)O
IUPAC Name (4-methylphenyl)boronic acid
Molecular Weight (g/mol) 135.96
PubChem CID 79799
Formula Weight 135.96
Percent Purity 98%
Chemical Name or Material 4-Methylbenzeneboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methylbenzeneboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 4-Methylbenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.