missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzeneboronic Acid, TRC

Product Code. 30467188
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30467188 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30467188 Supplier Toronto Research Chemicals Supplier No. TRCM28886010G
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methylbenzeneboronic Acid
CAS 5720-05-8
Molecular Formula C7H9BO2
Quantity 10 g
Synonym Boronic acid, B-(4-methylphenyl)-Boronic acid, (4-methylphenyl)- (9CI), p-Tolueneboronic acid (7CI,8CI), (4-Methylphenyl)boronic acid, (p-Methylphenyl)boronic acid, 4-Tolueneboronic acid, 4-Tolylboronic acid, NSC 62870, p-Methylbenzeneboronic acid, p-Tolylboronic acid
InChI Formula InChI=1S/C7H9BO2/c1-6-2-4-7(5-3-6)8(9)10/h2-5,9-10H,1H3
SMILES Cc1ccc(cc1)B(O)O
IUPAC Name (4-methylphenyl)boronic acid
Molecular Weight (g/mol) 135.96
Formula Weight 136.0696
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.