missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzenesulfonic Acid Monohydrate, TRC

Product Code. 30478186
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478186 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478186 Supplier Toronto Research Chemicals Supplier No. TRCM288740250G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methylbenzenesulphonic Acid Hydrate
CAS 6192-52-5
Purity Grade Notes HPLC
Molecular Formula C7 H8 O3 S . H2 O
Quantity 250 g
Synonym 4-Methylbenzenesulphonic acid hydrate, Benzenesulfonic acid, 4-methyl-, hydrate (1:1), Benzenesulfonic acid, 4-methyl-, monohydrate (9CI), p-Toluenesulfonic acid, monohydrate (8CI), 4-Methylbenzenesulfonic acid monohydrate, 4-Toluenesulfonic acid monohydrate, Tosic acid monohydrate, p-Toluenesulfonic acid monohydrate
InChI Formula InChI=1S/C7H8O3S.H2O/c1-6-2-4-7(5-3-6)11(8,9)10;/h2-5H,1H3,(H,8,9,10);1H2
SMILES O.Cc1ccc(cc1)S(=O)(=O)O
IUPAC Name 4-methylbenzenesulfonic acid;hydrate
Molecular Weight (g/mol) 190.22
Formula Weight 190.03
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.