missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzyl Alcohol, TRC

Product Code. 30473100
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30473100 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30473100 Supplier Toronto Research Chemicals Supplier No. TRCM80100550G
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methylbenzyl Alcohol
CAS 589-18-4
Molecular Formula C8 H10 O
Quantity 50 g
Synonym 4-(Hydroxymethyl)toluene, 4-Methylbenzyl alcohol, p-Methylbenzyl alcohol, p-Tolylcarbinol
InChI Formula InChI=1S/C8H10O/c1-7-2-4-8(6-9)5-3-7/h2-5,9H,6H2,1H3
SMILES Cc1ccc(CO)cc1
IUPAC Name p-tolylmethanol
Molecular Weight (g/mol) 122.16
Formula Weight 122.0732
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.