missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzyl isothiocyanate, 96%

Product Code. 11397576
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397576 10 g 10g
11387576 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397576 Supplier Thermo Scientific Alfa Aesar Supplier No. L11135.09
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 3694-46-0 | C9H9NS | 163.238 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3694-46-0
Molecular Formula C9H9NS
Molecular Weight (g/mol) 163.238
MDL Number MFCD00041118
InChI Key OAXIUBJXQISJEV-UHFFFAOYSA-N
Synonym 4-methylbenzyl isothiocyanate, 1-isothiocyanatomethyl-4-methylbenzene, p-methylbenzyl isothiocyanate, benzene, 1-isothiocyanatomethyl-4-methyl, isothiocyanic acid, p-methylbenzyl ester, benzene, 1-isothiocyanatomethyl-4-methyl-9ci, acmc-1cobu, 4-methylbenzylisothiocyanate, wln: scn1r d1, 4-12-00-02578 beilstein handbook reference
PubChem CID 19409
IUPAC Name 1-(isothiocyanatomethyl)-4-methylbenzene
SMILES CC1=CC=C(C=C1)CN=C=S

Specifications

Density 1.08
Boiling Point 97°C (1 mmHg)
Odor Pungent
Refractive Index 1.589
Quantity 10 g
UN Number UN2922
Beilstein 2082478
Sensitivity Moisture sensitive
Formula Weight 163.24
Percent Purity 96%
Chemical Name or Material 4-Methylbenzyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.