missing translation for 'onlineSavingsMsg'
Learn More

4'-Methylbiphenyl-2-carbonitrile, TRC

Product Code. 30479394
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30479394 100 g 100g
30470279 5 g 5g
30471480 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30479394 Supplier Toronto Research Chemicals Supplier No. TRCM294115100G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114772-53-1
Molecular Formula C14 H11 N
Molecular Weight (g/mol) 193.24
Synonym [1,1'-Biphenyl]-2-carbonitrile, 4'-methyl-, 4'-Methyl[1,1'-biphenyl]-2-carbonitrile, 2-(4-Methylphenyl)benzonitrile, 2-(p-Tolyl)benzonitrile, 2-Cyano-4'-methyl-1,1'-biphenyl, 2-Cyano-4'-methylbiphenyl, 2'-Cyano-4-methylbiphenyl, 4-Methyl-2'-cyanobiphenyl, 4'-Methyl-1,1'-biphenyl-2-ylcarbonitrile, 4'-Methyl-2-cyanobiphenyl, 4'-Methylbiphenyl-2-carbonitrile
IUPAC Name 2-(4-methylphenyl)benzonitrile
SMILES Cc1ccc(cc1)c2ccccc2C#N

Specifications

Quantity 100 g
InChI Formula InChI=1S/C14H11N/c1-11-6-8-12(9-7-11)14-5-3-2-4-13(14)10-15/h2-9H,1H3
Formula Weight 193.0891
Chemical Name or Material 4'-Methylbiphenyl-2-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.