missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbiphenyl, 98%

Product Code. 11457716
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11457716 1 g 1g
11467716 5 g 5g
11477716 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11457716 Supplier Thermo Scientific Alfa Aesar Supplier No. A14702.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 644-08-6 | C13H12 | 168.24 g/mol

4-Methylbiphenyl, is used as a flavoring ingredient in food industry. It is also used as an intermediate in agrochemcial, dyestuff, flavor and fragrance, pharmaceutical, syntheses material. It is also used as an OLED intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylbiphenyl, is used as a flavoring ingredient in food industry. It is also used as an intermediate in agrochemcial, dyestuff, flavor and fragrance, pharmaceutical, syntheses material. It is also used as an OLED intermediate.

Solubility
Insoluble in water. Soluble in alcohol and ether, very soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 644-08-6
Molecular Formula C13H12
Molecular Weight (g/mol) 168.24
MDL Number MFCD00008544
InChI Key ZZLCFHIKESPLTH-UHFFFAOYSA-N
Synonym 4-methylbiphenyl, 4-phenyltoluene, 4-methyl-1,1'-biphenyl, p-methylbiphenyl, p-methyldiphenyl, 4-methyldiphenyl, 1,1'-biphenyl, 4-methyl, biphenyl, 4-methyl, p-phenyltoluene, 4-methylphenyl benzene
PubChem CID 12566
SMILES CC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 44°C to 48°C
Density 1.015
Boiling Point 267°C to 268°C
Flash Point >110°C (230°F)
Quantity 1 g
Beilstein 1904468
Solubility Information Insoluble in water. Soluble in alcohol and ether,very soluble in organic solvents.
IUPAC Name 1-methyl-4-phenylbenzene
Formula Weight 168.24
Percent Purity 98%
Chemical Name or Material 4-Methylbiphenyl
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.