missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(Methylcarbamoyl)benzeneboronic acid, 98%

Product Code. 15419867
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15419867 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15419867 Supplier Alfa Aesar™ Supplier No. H53090.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 121177-82-0
Molecular Formula C8H10BNO3
Molecular Weight (g/mol) 178.982
MDL Number MFCD03788427
InChI Key UWKSYZHFTGONHY-UHFFFAOYSA-N
Synonym 4-n-methylaminocarbonyl phenylboronic acid, 4-methylcarbamoyl phenylboronic acid, 4-methylcarbamoyl phenyl boronic acid, 4-methylcarbamoyl benzeneboronic acid, boronic acid, 4-methylamino carbonyl phenyl, boronic acid,b-4-methylamino carbonyl phenyl, 4-methylamino carbonyl phenylboronic acid, 4-n-methylaminocarbonyl benzeneboronic acid, zlchem 836
PubChem CID 2773526
IUPAC Name [4-(methylcarbamoyl)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)C(=O)NC)(O)O

Specifications

Melting Point 166°C to 168°C
Quantity 1 g
Formula Weight 178.98
Percent Purity 98%
Chemical Name or Material 4-(Methylcarbamoyl)benzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.