missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Methylcyclohexene-1-boronic acid, 97%

Product Code. 15439497
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15439497 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15439497 Supplier Alfa Aesar™ Supplier No. H52759.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
4-Methylcyclohexene-1-boronic acid is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 850567-92-9
Melting Point 162°C to 166°C
Molecular Formula C7H13BO2
MDL Number MFCD06659857
Quantity 250 mg
Synonym 4-methyl-1-cyclohexen-1-ylboronic acid, 4-methylcyclohex-1-en-1-yl boronic acid, 4-methylcyclohex-1-en-1-ylboronic acid, 4-methylcyclohex-1-enylboronic acid, boronic acid, 4-methyl-1-cyclohexen-1-yl, boronic acid,b-4-methyl-1-cyclohexen-1-yl, acmc-209q0u, 4-methyl cyclohexen-1-ylboronic acid, 4-methylcyclohexen-1-yl boronic acid, 4-methyl-1-cyclohexenyl boronic acid
InChI Key SJZMIZIVYIOMIW-UHFFFAOYNA-N
SMILES CC1CCC(=CC1)B(O)O
IUPAC Name (4-methylcyclohexen-1-yl)boronic acid
Molecular Weight (g/mol) 139.99
PubChem CID 44119556
Formula Weight 139.99
Percent Purity 97%
Chemical Name or Material 4-Methylcyclohexene-1-boronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.