Learn More
Tocris Bioscience™ 4-Methylhistamine dihydrochloride
GREENER_CHOICE
Selective, high affinity H4 agonist
Brand:  Tocris Bioscience™ 2342/10
Encompass
Product Code. 13401074
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
4-Methylhistamine dihydrochloride is a potent, high affinity H4 receptor agonist (Ki = 7 nM) that displays > 100-fold selectivity over other human histamine receptor subtypes.
Specifications
Specifications
5-(2-Aminoethyl)-4-methylimidazole dihydrochloride
36376-47-3
10 mg
Soluble to 50mM in water