missing translation for 'onlineSavingsMsg'
Learn More

4-Methylhomopiperazine-4-aminobenzene trihydrochloride monohydrate, 90%, Thermo Scientific™

Product Code. 10516914
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10516914 250 mg Amber Glass Bottle 250mg
10482474 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10516914 Supplier Thermo Scientific Maybridge Supplier No. CC60364CB
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 913830-33-8
Molecular Formula C12H24Cl3N3O
Molecular Weight (g/mol) 332.694
MDL Number MFCD09025902
InChI Key RSGOMRQTIZQMIR-UHFFFAOYSA-N
Synonym 4-methylhomopiperazine-4-aminobenzene trihydrochloride monohydrate, 4-4-methyl-1,4-diazepan-1-yl aniline hydrate trihydrochloride, 4-4-methyl-1,4-diazepan-1-yl aniline trihydrochloride hydrate, 1-4-aminophenyl-4-methylhomopiperazine trihydrochloride hydrate, 4-4-methylhomopiperazin-1-yl aniline trihydrochloride monohydrate, 4-4-methyl-1,4-diazepan-1-yl aniline-hydrogen chloride-water 1/3/1
PubChem CID 44119580
IUPAC Name 4-(4-methyl-1,4-diazepan-1-yl)aniline;hydrate;trihydrochloride
SMILES CN1CCCN(CC1)C2=CC=C(C=C2)N.O.Cl.Cl.Cl

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Linear Formula Yes
Quantity 250 mg
Formula Weight 332.71
Percent Purity 90%
Chemical Name or Material 4-Methylhomopiperazine-4-aminobenzene trihydrochloride monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.