missing translation for 'onlineSavingsMsg'
Learn More

4-Methylpentan-2-One, Extra Pure, SLR, Fisher Chemical™

Product Code. 10427502 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
25 L
Packaging:
Amber glass bottle
Metal can
Plastic Coated Glass bottle
Unit Size:
1 L
2.5 L
25 L
5 L
500mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10427502 1 L Amber glass bottle 1 L
10356430 500 mL Amber glass bottle 500mL
10609113 500 mL Plastic Coated Glass bottle 500mL
10080270 2.5 L Amber glass bottle 2.5 L
10548060 2.5 L Plastic Coated Glass bottle 2.5 L
10469370 5 L Metal can 5 L
10528250 25 L Metal can 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 10427502 Supplier Fisher Chemical Supplier No. M/5078/15
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 108-10-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number 8938
InChI Key NTIZESTWPVYFNL-UHFFFAOYSA-N
Synonym 4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
PubChem CID 7909
ChEBI CHEBI:82344
IUPAC Name 4-methylpentan-2-one
SMILES CC(C)CC(=O)C

Specifications

Melting Point -84°C
Color Colorless
Boiling Point 117.4°C
Packaging Amber glass bottle
Quantity 1 L
Formula Weight 100.16g/mol
Physical Form Liquid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Methylpentan-2-one
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.