missing translation for 'onlineSavingsMsg'
Learn More

4-Methylphenyl 2,3,4,6-tetra-O-acetyl-beta-D-thiogalactopyranoside, Thermo Scientific Chemicals

Product Code. 15430497
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15430497 1 g 1g
15440497 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15430497 Supplier Thermo Scientific Alfa Aesar Supplier No. J66025.03
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28244-99-7
Molecular Formula C21H26O9S
Molecular Weight (g/mol) 454.49
MDL Number MFCD09750783
InChI Key XRFOIRMXQHXTRL-IFLJBQAJSA-N
PubChem CID 10366693
IUPAC Name [(2R,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-(4-methylphenyl)sulfanyloxan-2-yl]methyl acetate
SMILES CC1=CC=C(C=C1)SC2C(C(C(C(O2)COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 115°C to 116°C
Color White
Quantity 1 g
Synonym 4-methylphenyl 2,3,4,6-tetra-o-acetyl-b-d-thiogalactopyranoside, 4-methylphenyl 2,3,4,6-tetra-o-acetyl-1-thiohexopyranoside #, 4'-methylphenyl-1-thio-.beta.-d-galactoside, tetraacetate, 4-methylphenyl2,3,4,6-tetra-o-acetyl-b-d-thiogalactopyranoside, 4'-methylphenyl-1-thio-.beta.-d-galactoside, tetraacetate ester, 4-methylphenyl 2,3,4,6-tetra-o-acetyl-1-thio-beta-d-galactopyranoside, p-tolyl 2-o,3-o,4-o,6-o-tetraacetyl-1-thio-beta-d-galactopyranoside, 2r,3s,4s,5r,6s-2-acetoxymethyl-6-p-tolylthio tetrahydro-2h-pyran-3,4,5-triyl triacetate, 2r,3s,4s,5r,6s-3,4,5-tris acetyloxy-6-4-methylphenyl sulfanyl oxan-2-yl methyl acetate
Formula Weight 454.5
Physical Form Powder
Chemical Name or Material 4-Methylphenyl 2,3,4,6-tetra-O-acetyl-beta-D-thiogalactopyranoside

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.