missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methylpiperidine-2,2,6,6-d4, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 4-Methylpiperidine-2,2,6,6-D4 |
| CAS | 1219798-85-2 |
| Molecular Formula | C6 D4 H9 N |
| Quantity | 0.1 g |
| Synonym | Piperidine-2,2,6,6-d4, 4-methyl- (ACI), 4-Methylpiperidine-2,2,6,6-d4, 4-Pipecoline-2,2,6,6-d4, 4-Methylpiperidine-2,2,6,6-D4, 4-Methylpiperidine-d4 |
| InChI Formula | InChI=1 S/C6H13N/c1-6-2-4-7-5-3-6/h6-7 H,2-5H2,1H3/i4D2,5D2 |
| SMILES | [2 H]C1([2 H])CC(C)CC([2 H])([2 H])N1 |
| IUPAC Name | 2,2,6,6-tetradeuterio-4-methylpiperidine |
| Molecular Weight (g/mol) | 103.1988 |
| Formula Weight | 103.1299 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction