missing translation for 'onlineSavingsMsg'
Learn More

(4-Methylpiperidino)(2-piperidinyl)methanone hydrochloride, 95%, Thermo Scientific™

Product Code. 10593633
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10593633 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10593633 Supplier Thermo Scientific Maybridge Supplier No. MO01177DA
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 690634-80-1
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C12H23ClN2O
MDL Number MFCD06407988
Quantity 1 g
Synonym 4-methylpiperidino 2-piperidinyl methanone hydrochloride, 4-methyl-1-piperidinyl 2-piperidinyl methanone hydrochloride, 4-methyl-1-piperidine-2-carbonyl piperidine hydrochloride, 4-methylpiperidin-1-yl piperidin-2-yl methanone hydrochloride, 4-methylpiperidyl 2-piperidyl ketone, chloride, 4-methylpiperidin-1-yl-piperidin-2-ylmethanone hydrochloride, 4-methylpiperidin-1-yl piperidin-2-yl methanone-hydrogen chloride 1/1
InChI Key ITPHYLQZNJAUDV-UHFFFAOYSA-N
SMILES CC1CCN(CC1)C(=O)C2CCCCN2.Cl
IUPAC Name (4-methylpiperidin-1-yl)-piperidin-2-ylmethanone;hydrochloride
Molecular Weight (g/mol) 246.779
PubChem CID 2794697
Formula Weight 246.78
Percent Purity 95%
Chemical Name or Material (4-Methylpiperidino)(2-piperidinyl)methanone hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.