missing translation for 'onlineSavingsMsg'
Learn More

4'-Methylpropiophenone, 94%

Product Code. 11420257
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420257 100 g 100g
11430257 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11420257 Supplier Thermo Scientific Alfa Aesar Supplier No. B21673.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5337-93-9 | C10H12O | 148.21 g/mol

4?-Methylpropiophenone is a chemical reagent used in electrocarboxylation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Methylpropiophenone is a chemical reagent used in electrocarboxylation reactions.

Solubility
Soluble in chloroform and hexane.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5337-93-9
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.21
MDL Number MFCD00009312
InChI Key PATYHUUYADUHQS-UHFFFAOYSA-N
Synonym 4'-methylpropiophenone, 1-p-tolyl propan-1-one, p-methylpropiophenone, 4-methylpropiophenone, 1-4-methylphenyl propan-1-one, 1-propanone, 1-4-methylphenyl, p-tolyl ethyl ketone, p-methyl propiophenone, 1-p-tolylpropan-1-one, 1-4-methylphenyl-1-propanone
PubChem CID 21429
ChEBI CHEBI:59335
IUPAC Name 1-(4-methylphenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(C)C=C1

Specifications

Melting Point 7°C
Density 0.991
Boiling Point 238°C to 239°C
Flash Point 108°C (226°F)
Refractive Index 1.528
Quantity 100 g
Beilstein 2042137
Solubility Information Soluble in chloroform and hexane.
Formula Weight 148.21
Percent Purity 94%
Chemical Name or Material 4'-Methylpropiophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.