missing translation for 'onlineSavingsMsg'
Learn More

4′-Methylpropiophenone, TRC

Product Code. 30138722
Change view
Click to view available options
Quantity:
25 g
50 g
Unit Size:
25g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138722 50 g 50g
30136735 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138722 Supplier Toronto Research Chemicals Supplier No. TRCM33117550G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5337-93-9
Molecular Formula C10 H12 O
Molecular Weight (g/mol) 148.2
Synonym 4'-Methylpropiophenone, 1-(4-Methylphenyl)-1-propanone, 1-(4-Tolyl)-1-propanone, 4'-Methylpropiophenone, Ethyl 4-Methylphenyl Ketone, Ethyl p-Tolyl Ketone, NSC 852, p-Methylpropiophenone
IUPAC Name 1-(4-methylphenyl)propan-1-one
SMILES CCC(=O)c1ccc(C)cc1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C10H12O/c1-3-10(11)9-6-4-8(2)5-7-9/h4-7H,3H2,1-2H3
Formula Weight 148.0888
Chemical Name or Material 4'-Methylpropiophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.