missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylsulfonyl)benzenesulfonyl chloride, 95%, ACROS Organics™

Product Code. 10209380 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
2.5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10209380 2.5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10209380 Supplier Thermo Scientific Acros Supplier No. 424970025
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 82964-91-8
CAS Min % 94.0
CAS Max % 100.0
Melting Point 158°C to 163°C
Color Pink-Salmon to Yellow
Loss on Drying 5% max.
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C7H7ClO4S2
Linear Formula ClO2SC6H4SO2CH3
MDL Number MFCD00216486
Quantity 2.5 g
Synonym 4-methylsulfonyl benzenesulfonyl chloride, 4-methylsulfonyl benzene-1-sulfonyl chloride, 4-methanesulfonylbenzene-1-sulfonyl chloride, 4-methylsulphonyl benzenesulphonyl chloride, 4-methanesulfonylbenzenesulfonyl chloride, 4-methylsulfonylbenzenesulfonylchloride, pubchem5614, acmc-20amd2, chloro 4-methylsulfonyl phenyl sulfone, 4-methylsulfonyl phenylsulfonyl chloride
InChI Key TYJOQICPGZGYDT-UHFFFAOYSA-N
SMILES CS(=O)(=O)C1=CC=C(C=C1)S(Cl)(=O)=O
IUPAC Name 4-methanesulfonylbenzene-1-sulfonyl chloride
Molecular Weight (g/mol) 254.70
PubChem CID 2735182
Formula Weight 254.71
Percent Purity ≥94% (ex Cl)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Methylsulfonyl)benzenesulfonyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.