missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylsulfonyl)benzenesulfonyl chloride, 95%

Product Code. 11479400
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479400 1 g 1g
11489400 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11479400 Supplier Thermo Scientific Alfa Aesar Supplier No. H50264.03
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 82964-91-8 | C7H7ClO4S2 | 254.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82964-91-8
Molecular Formula C7H7ClO4S2
Molecular Weight (g/mol) 254.70
MDL Number MFCD00216486
InChI Key TYJOQICPGZGYDT-UHFFFAOYSA-N
Synonym 4-methylsulfonyl benzenesulfonyl chloride, 4-methylsulfonyl benzene-1-sulfonyl chloride, 4-methanesulfonylbenzene-1-sulfonyl chloride, 4-methylsulphonyl benzenesulphonyl chloride, 4-methanesulfonylbenzenesulfonyl chloride, 4-methylsulfonylbenzenesulfonylchloride, pubchem5614, acmc-20amd2, chloro 4-methylsulfonyl phenyl sulfone, 4-methylsulfonyl phenylsulfonyl chloride
PubChem CID 2735182
IUPAC Name 4-methylsulfonylbenzenesulfonyl chloride
SMILES CS(=O)(=O)C1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Melting Point 158°C to 163°C
Quantity 1 g
UN Number UN3261
Formula Weight 254.71
Percent Purity 95%
Chemical Name or Material 4-(Methylsulfonyl)benzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.