missing translation for 'onlineSavingsMsg'
Learn More

4-Methylthiazole-5-carboxaldehyde, 97%

Product Code. 15418168
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15418168 1 g 1g
15428168 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15418168 Supplier Thermo Scientific Alfa Aesar Supplier No. H61613.03
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 82294-70-0 | C5H5NOS | 127.161 g/mol

4-Methylthiazole-5-carboxaldehyde is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylthiazole-5-carboxaldehyde is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82294-70-0
Molecular Formula C5H5NOS
Molecular Weight (g/mol) 127.161
MDL Number MFCD07368277
InChI Key JJVIEMFQPALZOZ-UHFFFAOYSA-N
Synonym 4-methylthiazole-5-carbaldehyde, 4-methyl-5-thiazolecarboxaldehyde, 4-methylthiazole-5-carboxaldehyde, 4-methyl-5-formylthiazole, 4-methyl-5-thiazolecarbaldehyde, 5-formyl-4-methylthiazole, 4-methylthiazole-5-aldehyde, 4-methyl-5-thiazolyl aldehyde, 5-thiazolecarboxaldehyde, 4-methyl, 4-methyl-5-thiazole carboxaldehyde
PubChem CID 581339
IUPAC Name 4-methyl-1,3-thiazole-5-carbaldehyde
SMILES CC1=C(SC=N1)C=O

Specifications

Melting Point 74°C to 78°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 127.16
Percent Purity 97%
Chemical Name or Material 4-Methylthiazole-5-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.