missing translation for 'onlineSavingsMsg'
Learn More

4-Methylthiazole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10262502
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10262502 10 g 10g
10170642 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10262502 Supplier Thermo Scientific Maybridge Supplier No. MO00777EA
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 20485-41-0
Molecular Formula C5H5NO2S
Molecular Weight (g/mol) 143.16
MDL Number MFCD00626872
InChI Key ZGWGSEUMABQEMD-UHFFFAOYSA-N
Synonym 4-methylthiazole-5-carboxylic acid, 4-methyl-5-thiazolecarboxylic acid, 5-thiazolecarboxylic acid, 4-methyl, 4-methyl-thiazole-5-carboxylic acid, 4-methyl-5-thiazole carboxylic acid, 4-methylthiazole-5-carboxylicacid, pubchem8416, acmc-209far, 4-27-00-04008 beilstein handbook reference, ksc201q4f
PubChem CID 209805
IUPAC Name 4-methyl-1,3-thiazole-5-carboxylic acid
SMILES CC1=C(SC=N1)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 143.17
Percent Purity 97%
Chemical Name or Material 4-Methylthiazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.