missing translation for 'onlineSavingsMsg'
Learn More

4-Methylthio-2-butanone, 98%

Product Code. 11495826
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11495826 5 g 5g
11405836 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11495826 Supplier Thermo Scientific Alfa Aesar Supplier No. B22206.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 34047-39-7 | C5H10OS | 118.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34047-39-7
Molecular Formula C5H10OS
Molecular Weight (g/mol) 118.194
MDL Number MFCD00040029
InChI Key DRGHCRKOWMAZAO-UHFFFAOYSA-N
Synonym 4-methylthio-2-butanone, 2-butanone, 4-methylthio, 4-methylthio butan-2-one, unii-5703w1o1ab, 4-methylsulfanyl butan-2-one, potato butanone, fema no. 3375, 5-thiahexane-2-one, acmc-1afub
PubChem CID 61922
IUPAC Name 4-methylsulfanylbutan-2-one
SMILES CC(=O)CCSC

Specifications

Density 1.003
Boiling Point 105°C to 107°C (55 mmHg)
Flash Point 70°C (158°F)
Refractive Index 1.473
Quantity 5 g
Formula Weight 118.2
Percent Purity 98%
Chemical Name or Material 4-Methylthio-2-butanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.