missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)-6-(2-pyridyl)-1,3,5-triazin-2-amine, 97%, Thermo Scientific™

Product Code. 10225472
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10225472 1 g Amber Glass Bottle 1g
10573242 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10225472 Supplier Thermo Scientific Maybridge Supplier No. RF02651DA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 175204-53-2
Molecular Formula C9H9N5S
Molecular Weight (g/mol) 219.27
MDL Number MFCD00052773
InChI Key MAFCMTDUOYKYOR-UHFFFAOYSA-N
Synonym 4-methylthio-6-2-pyridyl-1,3,5-triazin-2-amine, 4-methylsulfanyl-6-pyridin-2-yl-1,3,5-triazin-2-amine, maybridge1_006429, 4-methylthio-6-2-pyridyl-1,3,5-triazine-2-ylamine, 1,3,5-triazin-2-amine,4-methylthio-6-2-pyridinyl, 4-methylthio-6-pyridin-2-yl-1,3,5-triazin-2-amine
PubChem CID 2782084
IUPAC Name 4-(methylsulfanyl)-6-(pyridin-2-yl)-1,3,5-triazin-2-amine
SMILES CSC1=NC(=NC(N)=N1)C1=CC=CC=N1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 219.27
Percent Purity 97%
Chemical Name or Material 4-(Methylthio)-6-(2-pyridyl)-1, 3, 5-triazin-2-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.