missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)-6-pyrazin-2-yl-1,3,5-triazin-2-amine, 97%, Thermo Scientific™

Product Code. 10790684
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10790684 1 g 1g
10700694 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10790684 Supplier Thermo Scientific Maybridge Supplier No. KM05093DA
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 175202-92-3
Molecular Formula C8H8N6S
Molecular Weight (g/mol) 220.254
MDL Number MFCD00068003
InChI Key SQOSJKISJKSCMM-UHFFFAOYSA-N
Synonym 4-methylthio-6-pyrazin-2-yl-1,3,5-triazin-2-amine, 4-methylsulfanyl-6-pyrazin-2-yl-1,3,5-triazin-2-amine, maybridge1_004945, 4-methylthio-6-pyrazin-2-yl-1,3,5-triazine-2-ylamine, 2-amino-4-methylthio-6-2-pyrazinyl-1,3,5-triazine
PubChem CID 2735378
IUPAC Name 4-methylsulfanyl-6-pyrazin-2-yl-1,3,5-triazin-2-amine
SMILES CSC1=NC(=NC(=N1)N)C2=NC=CN=C2

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 220.26
Percent Purity 97%
Chemical Name or Material 4-(Methylthio)-6-pyrazin-2-yl-1, 3, 5-triazin-2-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.