missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)benzaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number p-7028231
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
3446-89-7
C8H8OS
152.211
MFCD00006948
QRVYABWJVXXOTN-UHFFFAOYSA-N
4-methylthio benzaldehyde, benzaldehyde, 4-methylthio, 4-methylsulfanyl benzaldehyde, 4-methylmercaptobenzaldehyde, p-methylthio benzaldehyde, p-methylthiobenzaldehyde, unii-1tvq11biy8, 4-aldehyde thioanisole, p-methylmercaptobenzaldehyde
76985
4-methylsulfanylbenzaldehyde
CSC1=CC=C(C=C1)C=O
This item is not returnable. View return policy
3446-89-7
C8H8OS
152.211
MFCD00006948
QRVYABWJVXXOTN-UHFFFAOYSA-N
4-methylthio benzaldehyde, benzaldehyde, 4-methylthio, 4-methylsulfanyl benzaldehyde, 4-methylmercaptobenzaldehyde, p-methylthio benzaldehyde, p-methylthiobenzaldehyde, unii-1tvq11biy8, 4-aldehyde thioanisole, p-methylmercaptobenzaldehyde
76985
4-methylsulfanylbenzaldehyde
CSC1=CC=C(C=C1)C=O

4-(Methylthio)benzaldehyde is a building block for the synthesis of various pharmaceutical and biologically active compounds. It can be used for the synthesis of sulfur-containing terpyridine ligands. It is the intermediate for the synthesis of a class of pyrrole derivatives showing analgesic/anti-inflammatory activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Methylthio)benzaldehyde is a building block for the synthesis of various pharmaceutical and biologically active compounds. It can be used for the synthesis of sulfur-containing terpyridine ligands. It is the intermediate for the synthesis of a class of pyrrole derivatives showing analgesic/anti-inflammatory activity.

Solubility
Not miscible in water.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 6°C
Density 1.144
Boiling Point 118°C to 121°C (5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.649
Quantity 25 g
Beilstein 636158
Sensitivity Air Sensitive
Solubility Information Not miscible in water.
Formula Weight 152.22
Percent Purity 97%
Chemical Name or Material 4-(Methylthio)benzaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'