missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)benzaldehyde, TRC

Product Code. 30466551
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30466551 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30466551 Supplier Toronto Research Chemicals Supplier No. TRCM33066010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(Methylthio)benzaldehyde
CAS 3446-89-7
Molecular Formula C8H8OS
Quantity 10 g
Synonym 4-(Methylmercapto)benzaldehyde, 4-Methanesulfanylbenzaldehyde, 4-Methanethiobenzaldehyde, 4-Methylsulfanylbenzaldehyde, Methyl 4-Formylphenyl thioether, p-(Methylthio)benzaldehyde, p-Methylthiobenzaldehyde
InChI Formula InChI=1S/C8H8OS/c1-10-8-4-2-7(6-9)3-5-8/h2-6H,1H3
SMILES CSc1ccc(C=O)cc1
IUPAC Name 4-methylsulfanylbenzaldehyde
Molecular Weight (g/mol) 152.21
Formula Weight 152.0296
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.