missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl-beta-D-galactopyranoside, 97+%

Product Code. 11454527
Change view
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
0.25g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454527 0.25 g 0.25g
11444527 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11454527 Supplier Thermo Scientific Alfa Aesar Supplier No. B21075.01
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6160-78-7 | C16H18O8 | 338.31 g/mol

It is a substrate used for fluorometric assay for β-galactosidase. It is used to detect E-coli n the fluorogenic medium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a substrate used for fluorometric assay for β-galactosidase. It is used to detect E-coli n the fluorogenic medium.

Solubility
Soluble in water.

Notes
Moisture sensitive. Store at -20°C. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6160-78-7
Molecular Formula C16H18O8
Molecular Weight (g/mol) 338.31
MDL Number MFCD00036773,MFCD00063694
InChI Key YUDPTGPSBJVHCN-DZQJYWQESA-N
Synonym 4-methylumbelliferyl-beta-d-galactopyranoside, 4-methylumbelliferyl-galactopyranoside, 4-methylumbelliferyl beta-d-galactoside, 4-methylumbelliferyl b-d-galactoside, muga, 4-methylumbelliferyl beta-galactoside, 7-beta-d-galactopyranosyloxy-4-methyl-2h-1-benzopyran-2-one, 4-methylumbelliferyl b-d-galactopyranoside, 4-methyl-7-2s,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-chromen-2-one, 4-methyl-7-2s,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy chromen-2-one
PubChem CID 93577
IUPAC Name 4-methyl-7-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one
SMILES CC1=CC(=O)OC2=CC(O[C@@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O)=CC=C12

Specifications

Melting Point ∼260°C (decomposition)
Quantity 0.25 g
Beilstein 94673
Sensitivity Light sensitive
Solubility Information Soluble in water.
Optical Rotation −64° (c=1 in Pyridine, 24h)
Formula Weight 338.32
Percent Purity 98%
Chemical Name or Material 4-Methylumbelliferyl-beta-D-galactopyranoside

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.