missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl beta-D-glucopyranoside, 99%

Product Code. 15418069
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15418069

Brand: Thermo Scientific Alfa Aesar J66630.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18997-57-4 | C16H18O8 | 338.312 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18997-57-4
Molecular Formula C16H18O8
Molecular Weight (g/mol) 338.312
MDL Number MFCD00036773
InChI Key YUDPTGPSBJVHCN-YMILTQATSA-N
Synonym 4-methylumbelliferyl, a-d-glucoside, glc1-b-4mu, 7-, a-d-glucopyranosyloxy-4-methyl-2h-1-benzopyran-2-one, 4-methylumbelliferyl glucopyranoside, 4-methyl-7-2s,4s,5s-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy chromen-2-one
PubChem CID 2733779
IUPAC Name 4-methyl-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one
SMILES CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(C(O3)CO)O)O)O

Specifications

Melting Point 210°C to 212°C
Color White
Quantity 1 g
Beilstein 94673
Solubility Information Soluble in DMF or water
Formula Weight 338.31
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 4-Methylumbelliferyl beta-D-glucopyranoside

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.