Learn More
Melford 4-Methylumbelliferyl-β-L-fucopyranoside
Product Code. 30018506
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity Unit Size
30018506 100 mg 100mg
1 options
Product Code. 30018506 Supplier Melford Supplier No. M651000.1
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Fluorescent substrate for β-L-Fucosidase.
Fluorescent substrate for β-L-Fucosidase.
  • 4-Methylumbelliferone <50ppm
Specifications
Quantity 100 mg