missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl-N-acetyl-β-D-galactosaminide hydrate, 98%, Thermo Scientific™

Product Code. 10707412 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity unitSize
10707412 250 mg 250mg
1 options
This item is not returnable. View return policy

Product Code. 10707412

Brand: Thermo Scientific Acros 337122500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 36476-29-6
CAS Min % 97.5
CAS Max % 100.0
Melting Point 190.0°C to 192.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C18H21NO8·xH2O
MDL Number MFCD03791283
Quantity 250 mg
Specific Rotation Condition − 45.00 (20.00°C c=0.3,pyridine)
Specific Rotation − 45.00
Solubility Information (1% in warm MeOH/water 1:1) Clear colorless to light yellow
InChI Key QCTHLCFVVACBSA-SOVHRIKKSA-M
SMILES CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(C(O3)CO)O)O)N=C(C)[O-]
IUPAC Name N-[(2S,3R,4R,5R,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-(4-methyl-2-oxochromen-7-yl)oxyoxan-3-yl]ethanimidate
Molecular Weight (g/mol) 379.37
PubChem CID 22842632
Formula Weight 379.37
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylumbelliferyl-N-acetyl-β-D-galactosaminide hydrate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.