missing translation for 'onlineSavingsMsg'
Learn More

4-Morpholineethanol, TRC

Product Code. 30469198
Change view
Click to view available options
Quantity:
1 g
5 g
100 g
Unit Size:
100g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30469198 100 g 100g
30483082 5 g 5g
30467984 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30469198 Supplier Toronto Research Chemicals Supplier No. TRCM723750100G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-40-2
Molecular Formula C6 H13 N O2
Molecular Weight (g/mol) 131.17
Synonym 4-Morpholineethanol, 2-(4-Morpholino)ethyl alcohol, 2-(4-Morpholinyl)-1-ethanol, 2-(4-Morpholinyl)ethanol, 2-Morpholinoethanol, 4-(2-Hydroxyethyl)morpholine, N-(2-Hydroxyethyl)morpholine, N-(β-Hydroxyethyl)morpholine, NSC 1946, β-Morpholinoethanol
IUPAC Name 2-morpholin-4-ylethanol
SMILES OCCN1CCOCC1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C6H13NO2/c8-4-1-7-2-5-9-6-3-7/h8H,1-6H2
Formula Weight 131.0946
Chemical Name or Material 4-Morpholineethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.