missing translation for 'onlineSavingsMsg'
Learn More

4-Morpholinepropionitrile, 98+%

Product Code. 11392557
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392557 25 g 25g
11382557 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392557 Supplier Thermo Scientific Alfa Aesar Supplier No. L11664.14
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4542-47-6 | C7H12N2O | 140.186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4542-47-6
Molecular Formula C7H12N2O
Molecular Weight (g/mol) 140.186
MDL Number MFCD00006178
InChI Key WXVKGHVDWWXBJX-UHFFFAOYSA-N
Synonym 3-morpholinopropionitrile, 3-morpholin-4-yl propanenitrile, 3-4-morpholino propionitrile, 4-morpholinepropanenitrile, cyanoethylmorpholine, 4-morpholinepropionitrile, beta-morpholinopropionitrile, n-2-cyanoethyl morpholine, propionitrile, 3-morpholino, 3-morpholinopropanenitrile
PubChem CID 78298
IUPAC Name 3-morpholin-4-ylpropanenitrile
SMILES C1COCCN1CCC#N

Specifications

Melting Point 20°C to 21°C
Density 1.037
Boiling Point 106°C to 109°C (7 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.472
Quantity 25 g
UN Number UN3276
Beilstein 110425
Formula Weight 140.19
Percent Purity ≥98%
Chemical Name or Material 4-Morpholinepropionitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.