missing translation for 'onlineSavingsMsg'
Learn More

4-Morpholinoaniline, TRC

Product Code. 30471583
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30471583 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30471583 Supplier Toronto Research Chemicals Supplier No. TRCM72374525G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Morpholinoaniline
CAS 2524-67-6
Molecular Formula C10 H14 N2 O
Quantity 25 g
Synonym Benzenamine','4-(4-morpholinyl)-, Morpholine','4-(p-aminophenyl)- (6CI,7CI,8CI), 4-(4-Morpholinyl)benzenamine, (4-(Morpholino)phenyl)amine, 4-(4-Aminophenyl)morpholine, 4-(4-Morpholinyl)aniline, 4-(Morpholin-4-yl)phenylamine, 4-Morpholinoaniline, 4-Morpholinobenzenamine, 4-N-Morpholinoaniline, N-(4-Aminophenyl)morpholine, N-(4'-Aminophenyl)morpholine, NSC 26334, p-Morpholinoaniline
InChI Formula InChI=1S/C10H14N2O/c11-9-1-3-10(4-2-9)12-5-7-13-8-6-12/h1-4H,5-8,11H2
SMILES Nc1ccc(cc1)N2CCOCC2
IUPAC Name 4-morpholin-4-ylaniline
Molecular Weight (g/mol) 178.231
Formula Weight 178.111
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.