missing translation for 'onlineSavingsMsg'
Learn More

4-Morpholinobenzonitrile, 97%, Thermo Scientific™

Product Code. 10767295
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10767295 5 g Amber Glass Bottle 5g
10547474 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10767295 Supplier Thermo Scientific Maybridge Supplier No. NRB01111DE
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 10282-31-2
Molecular Formula C11H12N2O
Molecular Weight (g/mol) 188.23
MDL Number MFCD00100115
InChI Key ZSCUWVQXQDCSRV-UHFFFAOYSA-N
PubChem CID 394808
IUPAC Name 4-morpholin-4-ylbenzonitrile
SMILES C1COCCN1C2=CC=C(C=C2)C#N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Synonym 4-morpholinobenzonitrile, 4-4-cyanophenyl morpholine, 4-morpholin-4-yl benzonitrile, 4-4-morpholinyl benzonitrile, 4-morpholin-4-yl-benzonitrile, benzonitrile, 4-4-morpholinyl, benzonitrile,4-4-morpholinyl, 4-morpholin-4-ylbenzenecarbonitrile, pubchem18708, acmc-1bqst
Formula Weight 188.23
Percent Purity 97%
Chemical Name or Material 4-Morpholinobenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.