missing translation for 'onlineSavingsMsg'
Learn More

4-Morpholinophenyl isothiocyanate, Thermo Scientific™

Product Code. 10126162
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10126162 1 g Amber Glass Bottle 1g
10157522 250 mg Amber Glass Bottle 250mg
10326544 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10126162 Supplier Thermo Scientific Maybridge Supplier No. CC17407DA
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 51317-66-9
Molecular Formula C11H12N2OS
Molecular Weight (g/mol) 220.29
InChI Key AXUXRZZYZBZQAR-UHFFFAOYSA-N
PubChem CID 224862
IUPAC Name 4-(4-isothiocyanatophenyl)morpholine
SMILES C1COCCN1C2=CC=C(C=C2)N=C=S

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 4-morpholinophenyl isothiocyanate, 4-4-isothiocyanatophenyl morpholine, 4-morpholinophenylisothiocyanate, morpholine, 4-4-isothiocyanatophenyl, 4-4-isothiocyanatophenyl morpholin, 4-morpholin-4-ylbenzenisothiocyanate, 4-4-morpholinyl phenyl isothiocyanate, 4-4-isothiocyanato-phenyl-morpholine, morpholine,4-4-isothiocyanatophenyl, 4-morpholin-4-yl phenyl isothiocyanate
Formula Weight 220.29
Percent Purity 97%
Chemical Name or Material 4-Morpholinophenyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.