missing translation for 'onlineSavingsMsg'
Learn More

4-(Morpholinosulfonyl)aniline, 97%, Thermo Scientific™

Product Code. 10529982
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10529982 10 g 10g
10345162 1 g 1g
10200255 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10529982 Supplier Thermo Scientific Maybridge Supplier No. TL00826EA
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 21626-70-0
Molecular Formula C10H14N2O3S
Molecular Weight (g/mol) 242.293
MDL Number MFCD00277213
InChI Key FTKHPQFFQRKOJC-UHFFFAOYSA-N
Synonym 4-morpholinosulfonyl aniline, 4-morpholin-4-ylsulfonyl aniline, 4-morpholine-4-sulfonyl aniline, compound 82 208, 4-morpholine-4-sulfonyl-phenylamine, 4-4-morpholinylsulfonyl aniline, compound 82/208, morpholine, 4-sulfanilyl, sulfanilic acid, morpholide, 4-aminophenyl morpholino sulfone
PubChem CID 210794
IUPAC Name 4-morpholin-4-ylsulfonylaniline
SMILES C1COCCN1S(=O)(=O)C2=CC=C(C=C2)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 242.3
Percent Purity 97%
Chemical Name or Material 4-(Morpholinosulfonyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.