missing translation for 'onlineSavingsMsg'
Learn More

4-(N-1-Dimethylaminonaphthalene-5-sulfonylamino)phenyl Isothiocyanate, TRC

Product Code. 30384623
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30384623 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384623 Supplier Toronto Research Chemicals Supplier No. TRCD461000100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material 4-(N-1-Dimethylaminonaphthalene-5-sulfonylamino)phenyl Isoth
CAS 102417-94-7
Molecular Formula C19 H17 N3 O2 S2
Quantity 100 mg
InChI Formula InChI=1S/C19H17N3O2S2/c1-22(2)18-7-3-6-17-16(18)5-4-8-19(17)26(23,24)21-15-11-9-14(10-12-15)20-13-25/h3-12,21H,1-2H3
SMILES CN(C)c1cccc2c(cccc12)S(=O)(=O)Nc3ccc(cc3)N=C=S
IUPAC Name 5-(dimethylamino)-N-(4-isothiocyanatophenyl)naphthalene-1-sulfonamide
Molecular Weight (g/mol) 383.49
Formula Weight 383.0762
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.