missing translation for 'onlineSavingsMsg'
Learn More

4-N-BOC-Amino-4-carboxytetrahydropyran, 95%

Product Code. 10590722 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
2 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10590722 500 mg Glass bottle 500mg
10359093 2 g Glass bottle 2g
11925421 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10590722 Supplier Thermo Scientific Acros Supplier No. 360465000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 172843-97-9 | C11H19NO5 | 245.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 172843-97-9
Molecular Formula C11H19NO5
Molecular Weight (g/mol) 245.27
MDL Number MFCD02683136
InChI Key SPPDKPRJPFTBEV-UHFFFAOYSA-N
Synonym 4-boc-amino tetrahydropyran-4-carboxylic acid, 4-n-boc-amino-4-carboxytetrahydropyran, 4-tert-butoxycarbonyl amino tetrahydro-2h-pyran-4-carboxylic acid, 4-tert-butoxycarbonyl amino oxane-4-carboxylic acid, 4-tert-butoxy carbonyl amino oxane-4-carboxylic acid, 4-tert-butoxycarbamoyl tetrahydropyran-4-carboxylic acid, 4-tert-butoxycarbonylamino tetrahydro-2h-pyran-4-carboxylic acid, 4-tert-butoxy carbonylamino-2h-3,4,5,6-tetrahydropyran-4-carboxylic acid
PubChem CID 1268219
IUPAC Name 4-[(2-methylpropan-2-yl)oxycarbonylamino]oxane-4-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1(CCOCC1)C(=O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 500 mg
Formula Weight 245.27
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 4-N-BOC-Amino-4-carboxytetrahydropyran, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 4-N-BOC-Amino-4-carboxytetrahydropyran
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.