missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butoxybenzeneboronic acid, 98%

Product Code. 15479637
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15479637 1 g 1g
15489637 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15479637 Supplier Thermo Scientific Alfa Aesar Supplier No. H52865.03
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 105365-51-3 | C10H15BO3 | 194.04 g/mol

Used as a reactant for Suzuki-Miyaura cross-coupling reactions, condensation reactions, synthesis of azobenzene-functionalized compounds. Also used for preparation of highly fluorescent diketopyrrolopyrrole derivatives, rhodium-catalyzed Suzuki-type cross-coupling and copper-catalyzed asymmetric conjugate reduction of coumarins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a reactant for Suzuki-Miyaura cross-coupling reactions, condensation reactions, synthesis of azobenzene-functionalized compounds. Also used for preparation of highly fluorescent diketopyrrolopyrrole derivatives, rhodium-catalyzed Suzuki-type cross-coupling and copper-catalyzed asymmetric conjugate reduction of coumarins.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105365-51-3
Molecular Formula C10H15BO3
Molecular Weight (g/mol) 194.04
MDL Number MFCD03427054
InChI Key QUPFQMXWFNJUNJ-UHFFFAOYSA-N
Synonym 4-n-butoxyphenylboronic acid, 4-butoxyphenyl boronic acid, 4-n-butoxyphenyl boronic acid, p-butoxyphenylboronic acid, 4-butyloxyphenylboronic acid, 4-n-butoxy benzeneboronic acid, 4-butoxymethylboronic acid, boronic acid, 4-butoxyphenyl, pubchem9549, 4-butoxybenzeneboronic acid
PubChem CID 3836310
IUPAC Name (4-butoxyphenyl)boronic acid
SMILES CCCCOC1=CC=C(C=C1)B(O)O

Specifications

Melting Point 102°C to 104°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 194.04
Percent Purity 98%
Chemical Name or Material 4-n-Butoxybenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.