missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butoxybenzenesulfonyl chloride, 97%

Product Code. 11357656
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357656 5 g 5g
11367656 250 mg 250mg
11347656 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11357656 Supplier Thermo Scientific Alfa Aesar Supplier No. L11258.06
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1138-56-3 | C10H13ClO3S | 248.721 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1138-56-3
Molecular Formula C10H13ClO3S
Molecular Weight (g/mol) 248.721
MDL Number MFCD00052344
InChI Key HGKWMUBXVMFXNC-UHFFFAOYSA-N
Synonym 4-n-butoxy benzenesulfonyl chloride, 4-butoxybenzene-1-sulfonyl chloride, 4-n-butoxy benzenesulphonyl chloride, benzenesulfonyl chloride, 4-butoxy, 4-butoxy-benzenesulfonyl chloride, 4-n-butoxybenzenesulfonyl chloride, 4-butoxybenzenesulfonylchloride, pubchem5072, 4-n-butoxy benzenes, buttpark 9650-54
PubChem CID 517990
IUPAC Name 4-butoxybenzenesulfonyl chloride
SMILES CCCCOC1=CC=C(C=C1)S(=O)(=O)Cl

Specifications

Boiling Point 130°C to 131°C (0.6 mmHg)
Refractive Index 1.542
Quantity 5 g
UN Number UN3265
Beilstein 1109264
Sensitivity Moisture sensitive
Formula Weight 248.73
Percent Purity 97%
Chemical Name or Material 4-n-Butoxybenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.