missing translation for 'onlineSavingsMsg'
Learn More

4-(n-Butoxycarbonyl)phenyl isocyanate, 99%, Thermo Scientific™

Product Code. 11392886
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11392886 1 g 1g
11302896 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392886 Supplier Thermo Scientific Alfa Aesar Supplier No. L10606.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102561-47-7
Molecular Formula C12H13NO3
Molecular Weight (g/mol) 219.24
MDL Number MFCD00037043
InChI Key PVCGQSQNUXNTNA-UHFFFAOYSA-N
Synonym 4-n-butoxycarbonyl phenyl isocyanate, acmc-20e5xd, 4-carbobutoxy phenyl isocyanate, butyl 4-isocyanatobenzoate, 4-n-butoxycarbonyl phenylisocyanate, 4-carbobutoxyphenyl isocyanate, 4-n-carbobutoxyphenyl isocyanate, benzoic acid,4-isocyanato-, butyl ester, benzoic acid, 4-isocyanato-, butyl ester 9ci, 4-n-butoxycarbonyl phenyl isocyanate; 4-carbobutoxy phenyl isocyanate
PubChem CID 4112112
IUPAC Name butyl 4-isocyanatobenzoate
SMILES CCCCOC(=O)C1=CC=C(C=C1)N=C=O

Specifications

Density 1.105
Boiling Point 295°C
Flash Point >110°C (230°F)
Refractive Index 1.5275
Quantity 1 g
UN Number UN2206
Beilstein 7640773
Sensitivity Moisture sensitive
Formula Weight 219.24
Percent Purity 99%
Chemical Name or Material 4-(n-Butoxycarbonyl)phenyl isocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.