missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butoxyphenyl isocyanate, 98%

Product Code. 11303426
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11303426

Brand: Thermo Scientific Alfa Aesar L12877.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 28439-86-3 | C11H13NO2 | 191.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28439-86-3
Molecular Formula C11H13NO2
Molecular Weight (g/mol) 191.23
MDL Number MFCD00037042
InChI Key ASFIDVOMDQBCNP-UHFFFAOYSA-N
Synonym 4-n-butoxyphenyl isocyanate, 4-butoxyphenyl isocyanate, benzene, 1-butoxy-4-isocyanato, 1-butoxy-4-isocyanato-benzene, pubchem16232, 4-butoxybenzenisocyanate
PubChem CID 2769629
IUPAC Name 1-butoxy-4-isocyanatobenzene
SMILES CCCCOC1=CC=C(C=C1)N=C=O

Specifications

Density 1.055
Boiling Point 149°C to 150°C (3 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.522
Quantity 1 g
UN Number UN2206
Beilstein 2107851
Sensitivity Moisture sensitive
Formula Weight 191.23
Percent Purity 98%
Chemical Name or Material 4-n-Butoxyphenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.