missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butoxyphenyl isocyanate, 98%

Product Code. 11313426
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11313426 5 g 5g
11303426 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11313426 Supplier Thermo Scientific Alfa Aesar Supplier No. L12877.06
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 28439-86-3 | C11H13NO2 | 191.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28439-86-3
Molecular Formula C11H13NO2
Molecular Weight (g/mol) 191.23
MDL Number MFCD00037042
InChI Key ASFIDVOMDQBCNP-UHFFFAOYSA-N
Synonym 4-n-butoxyphenyl isocyanate, 4-butoxyphenyl isocyanate, benzene, 1-butoxy-4-isocyanato, 1-butoxy-4-isocyanato-benzene, pubchem16232, 4-butoxybenzenisocyanate
PubChem CID 2769629
IUPAC Name 1-butoxy-4-isocyanatobenzene
SMILES CCCCOC1=CC=C(C=C1)N=C=O

Specifications

Density 1.055
Boiling Point 149°C to 150°C (3 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.522
Quantity 5 g
UN Number UN2206
Beilstein 2107851
Sensitivity Moisture sensitive
Formula Weight 191.23
Percent Purity 98%
Chemical Name or Material 4-n-Butoxyphenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.