missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butylbenzenesulfonyl chloride, 97%

Product Code. 11417907
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11417907 5 g 5g
11427907 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11417907 Supplier Thermo Scientific Alfa Aesar Supplier No. A16577.06
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 54997-92-1 | C10H13ClO2S | 232.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54997-92-1
Molecular Formula C10H13ClO2S
Molecular Weight (g/mol) 232.72
MDL Number MFCD00173908
InChI Key OVFZELSNOHIDEF-UHFFFAOYSA-N
Synonym 4-butylbenzene-1-sulfonyl chloride, 4-n-butylbenzenesulfonyl chloride, 4-n-butyl benzenesulphonyl chloride, 4-butyl-benzenesulfonyl chloride, 4-n-butylphenylsulfonyl chloride, 4-n-butylbenzenesulphonyl chloride, benzenesulfonyl chloride, 4-butyl, 4-butylphenyl chlorosulfone, pubchem5484, 4-n-butylbenzenesulfonylchloride
PubChem CID 2737352
IUPAC Name 4-butylbenzenesulfonyl chloride
SMILES CCCCC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Melting Point 13°C to 15°C
Boiling Point 126°C to 128°C (0.6 mmHg)
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 232.73
Percent Purity 97%
Chemical Name or Material 4-n-Butylbenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.