missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butylbenzoyl chloride, 98%

Product Code. 11447387
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11447387 5 g 5g
11457387 25 g 25g
11467387 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11447387 Supplier Thermo Scientific Alfa Aesar Supplier No. A15082.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 28788-62-7 | C11H13ClO | 196.67 g/mol

4-Butylbenzoyl chloride was used in the synthesis of 2,5-bis[(4-butylbenzoyl)oxy]styrene (BBOS, monomer).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Butylbenzoyl chloride was used in the synthesis of 2,5-bis[(4-butylbenzoyl)oxy]styrene (BBOS, monomer).

Solubility
Reacts with water.

Notes
Moisture Sensitive, store away from moisture. Store in cool, dry place in tightly closed container. Store away from oxidizing agent. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28788-62-7
Molecular Formula C11H13ClO
Molecular Weight (g/mol) 196.67
MDL Number MFCD00000699
InChI Key OUOWCSJYDCPVDM-UHFFFAOYSA-N
Synonym 4-n-butylbenzoyl chloride, benzoyl chloride, 4-butyl, p-butylbenzoyl chloride, 4-n-butyl-benzoyl chloride, 4-butylbenzoylchloride, 4-n-butylbenzoylchloride, p-n-butyl benzoylchloride, p-n-butylbenzoyl chloride, 4-1-butylbenzoyl chloride, acmc-209h4o
PubChem CID 120049
IUPAC Name 4-butylbenzoyl chloride
SMILES CCCCC1=CC=C(C=C1)C(Cl)=O

Specifications

Boiling Point 145°C to 147°C (11 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3265
Formula Weight 196.68
Percent Purity 98%
Chemical Name or Material 4-n-Butylbenzoyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.