missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-n-Decylbenzoyl chloride, 98%
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Description
Employed as a pharmaceutical and chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Employed as a pharmaceutical and chemical intermediate.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Store away from oxidizing agents, water/moisture. Store under inert gas.
Employed as a pharmaceutical and chemical intermediate.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Store away from oxidizing agents, water/moisture. Store under inert gas.
Specifications
Specifications
| Boiling Point | 171°C to 173°C (2 mmHg) |
| Flash Point | >110°C (230°F) |
| Quantity | 25 g |
| UN Number | UN3265 |
| Sensitivity | Moisture sensitive |
| Synonym | 4-n-decylbenzoyl chloride, benzoyl chloride, 4-decyl, p-decylbenzoyl chloride, 4-decylbenzoylchloride, p-n-decylbenzoyl chloride, benzoyl chloride,4-decyl, 4-n-decyl benzoyl chloride, 4-decylbenzoic acid chloride |
| Solubility Information | Reacts with water. |
| Formula Weight | 280.84 |
| Percent Purity | 98% |
| Chemical Name or Material | 4-n-Decylbenzoyl chloride |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction