missing translation for 'onlineSavingsMsg'
Learn More

4-(N,N-Dimethylamino)phenylboronic acid, 96%

Product Code. 10068841 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10068841 500 mg Glass bottle 500mg
10045713 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10068841 Supplier Thermo Scientific Acros Supplier No. 359235000
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 28611-39-4 | C8H12BNO2 | 165.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28611-39-4
Molecular Formula C8H12BNO2
Molecular Weight (g/mol) 165.00
MDL Number MFCD01074642
InChI Key RIIPFHVHLXPMHQ-UHFFFAOYSA-N
Synonym 4-dimethylamino phenylboronic acid, 4-dimethylamino phenyl boronic acid, 4-n,n-dimethylamino phenylboronic acid, 4-dimethylamino benzeneboronic acid, 4-dimethylaminophenylboronic acid, 4-dimethylaminophenyl boronic acid, 4-dimethylamino phenyl boranediol, 4-n,n-dimethylphenylboronic acid
PubChem CID 2734344
IUPAC Name [4-(dimethylamino)phenyl]boronic acid
SMILES CN(C)C1=CC=C(C=C1)B(O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 227°C
Color Blue-Gray or Gray-Green to White
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Quantity 500 mg
Formula Weight 165
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 4-(N, N-Dimethylamino)phenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-(N,N-Dimethylamino)phenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.