missing translation for 'onlineSavingsMsg'
Learn More

4-n-Pentylphenylacetylene, 97%

Product Code. 15467418
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
Product Code. Quantity unitSize
15467418 2 g 2g
15477418 10 g 10g
2 options
This item is not returnable. View return policy

Product Code. 15467418

Brand: Thermo Scientific Alfa Aesar H60913.04

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 79887-10-8 | C13H16 | 172.27 g/mol

4-n-Pentylphenylacetylene is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-n-Pentylphenylacetylene is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79887-10-8
Molecular Formula C13H16
Molecular Weight (g/mol) 172.27
MDL Number MFCD00173884
InChI Key APGNXGIUUTWIRE-UHFFFAOYSA-N
Synonym 4-n-pentylphenylacetylene, 4-pentylphenylacetylene, 1-ethynyl-4-pentyl-benzene, 1-eth-1-ynyl-4-pentylbenzene, benzene, 1-ethynyl-4-pentyl, 4-amylphenylacetylene, 4-ethynylpentylbenzene, acmc-209pid, 4-pentylphenyl acetylene
PubChem CID 2775131
IUPAC Name 1-ethynyl-4-pentylbenzene
SMILES CCCCCC1=CC=C(C=C1)C#C

Specifications

Density 0.885
Boiling Point 172°C
Flash Point 103°C (217°F)
Refractive Index 1.523
Quantity 2 g
Formula Weight 172.27
Percent Purity 97%
Chemical Name or Material 4-n-Pentylphenylacetylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.